Learn More
Waters X3 CORTECS Shield RP18 Column, 90A, 2.7 um, 3 mm X 30 mm
Brand:  Waters 176003917
Encompass
X3 CORTECS Shield RP18 Column, 90A, 2.7 um, 3 mm X 30 mm
Product Code. 17132782
  • € 3750.00 / Pack of 3