Learn More
WLD Tec X6 Express 445 gas cartridges .
Brand:  WLD Tec 6.000.186
X6 Express 445 gas cartridges .
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 17582095
  • ONLINE EXCLUSIVE PRICE
    € 99.75
    € 53.37 / Pack of 6
    Save € 46.38
    46% Off
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Express 445 gas cartridges
Signal Word
  • Danger
Hazard Category
  • Flammable gas.
  • Gases under pressure
Hazard Statement
  • H220-Extremely flammable gas.
  • H280-Contains gas under pressure; may explode if heated.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P377-Leaking gas fire: Do not extinguish, unless leak can be stopped safely.
  • P381-Eliminate all ignition sources if safe to do so.
  • P410+P403-Protect from sunlight Store in a well-ventilated place.
Supplemental information
  • MIXTURE LIST-Contains : Hydrocarbons, C3-4-rich, petroleum distillate, propene