missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse C-Reactive Protein/CRP Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
Description
Includes
Mouse CRP Microplate; Mouse CRP Standard; Mouse CRP Control; Mouse CRP Conjugate; Assay Diluent RD1W; Calibrator Diluent RD5P Concentrate; Wash Buffer Concentrate; Color Reagent A; Color Reagent B; Stop Solution; Plate Sealers
The Mouse C-Reactive Protein/CRP Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse C-Reactive Protein/CRP in cell culture supernates (50 ul), tissue lysates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
Specifications
Specifications
| Assay Range | 0.16 - 10ng/mL |
| Assay Sensitivity | 0.015ng/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Lysates |
| Specificity | Natural and Recombinant Mouse CRP |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Sample Volume | Cell Culture Supernates (50uL),Tissue Lysates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction