missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XAB1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97600UV
This item is not returnable.
View return policy
Description
XAB1 Polyclonal specifically detects XAB1 in Human samples. It is validated for Western Blot.
Specifications
| XAB1 | |
| Polyclonal | |
| Western Blot | |
| ATP(GTP)-binding protein, ATPBD1A, GPN-loop GTPase 1, GTPase, MBD2-interacting protein, MBDin, MBDINXPA binding protein 1, XAB1FLJ51176, XPA-binding protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human XAB1 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 11321 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction