missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XAB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565527
This item is not returnable.
View return policy
Description
Immunogen sequence: RAQVKHRCVTD PAYEDVNNCH ERAFVFMHKM PRLWLDYCQF LMDQGRVTHT RRTFDRALRA LPITQHSRIW PLYLRFLRSH PLPETAVRGY RRFLKLSPES AEEYIEYLKS SDRLD Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
A novel protein designated XAB2 (XPA-binding protein 2) interacts with XPA, a factor central to both nucleotide excision repair subpathways. Data suggests that XAB2 is a novel component involved in transcription-coupled repair and transcription.
Specifications
| XAB2 | |
| Polyclonal | |
| Unconjugated | |
| XAB2 | |
| 0610041O14Rik; Adapter protein ATH-55; Ath55; AV025587; crn-related protein kim1; DKFZp762C1015; HCNP; HCNP protein; HCRN; KIAA1177; NTC90; PP3898; Pre-mRNA-splicing factor SYF1; Protein HCNP; SYF1; SYF1 homolog, RNA splicing factor; SYF1 pre-mRNA-splicing factor; Xab2; XPA binding protein 2; XPA-binding protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56949 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HCS7 | |
| XAB2 | |
| Recombinant Human XAB2. Recombinant protein control fragment (Product #RP-106201). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto