missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ XAGE1 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Specifications
Specifications
| Gene ID (Entrez) | 653067 |
| Species | Human |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-262 |
| Gene Alias | CT12.1, CTP9, GAGED2, member 1 c, protein XAGE-1, x antigen family, member 1 C |
| Gene Symbol | XAGE1B |
| Quantity | 0.1 mg |
| Research Category | Apoptosis, Cancer, Cellular Markers, Signal Transduction, Tumor Suppressors |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur