Learn More
Xanthine Oxidase Antibody (8A9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26250-50ul
 View more versions of this product
Product Code. 30032440
  • € 355.00 / 50µL
Description
Xanthine Oxidase Monoclonal antibody specifically detects Xanthine Oxidase in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Xanthine Oxidase
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA, Immunohistochemistry
8A9
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
EC 1.17.1.4, EC 1.7.2.2, xanthene dehydrogenase, xanthine dehydrogenase, xanthine dehydrogenase/oxidase, xanthine oxidase, xanthine oxidoreductase, XDHA, XO, XOR
A synthesized peptide derived from Human Xanthine Oxidase [UniProt P47989]
50 μL
Signal Transduction
7498
Store at -20 to -70C. Avoid freeze-thaw cycles.