missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XBP1 Antibody (525904), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4257K
This item is not returnable.
View return policy
Description
XBP1 Monoclonal antibody specifically detects XBP1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| XBP1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 525904 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2Tax-responsive element-binding protein 5 | |
| E. coli-derived recombinant human XBP1S, Ser188-Leu287, Accession # NP_001073007 | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
| 7494 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction