missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XBP1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77252AF647
This item is not returnable.
View return policy
Description
XBP1 Polyclonal antibody specifically detects XBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| XBP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 647 | |
| TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2Tax-responsive element-binding protein 5 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human XBP-1. The immunogen is located within the last 50 amino acids of XBP-1. Amino Acid Squence: LPAWRSSQRSTQKDPVPY | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
| 7494 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction