missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XBP1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77253FR
This item is not returnable.
View return policy
Description
XBP1 Polyclonal antibody specifically detects XBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| XBP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2Tax-responsive element-binding protein 5 | |
| Antibody was raised against a peptide corresponding to 17 amino acids near the amino terminus of human XBP1. The immunogen is located within amino acids 40 - 90 of XBP1. Amino Acid Sequence: GLPQARKRQRLTHLSPE | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
| 7494 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction