missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation XBridge BEH300 C18 Column

Product Code. 13115229
Encompass
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
4.6 mm
Particle Size:
3.5 μm
5 μm
10 μm
Unit Size:
Each
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size unitSize
13115229 50 mm 4.6 mm 3.5 μm Each
13135229 150 mm 4.6 mm 3.5 μm Each
13175219 150 mm 1 mm 3.5 μm Each
13145229 250 mm 4.6 mm 3.5 μm Each
13155219 50 mm 1 mm 3.5 μm Each
13165219 100 mm 1 mm 3.5 μm Each
13185219 50 mm 2.1 mm 3.5 μm Each
10657725 100 mm 2.1 mm 3.5 μm Each
13195219 150 mm 2.1 mm 3.5 μm Each
13105229 250 mm 2.1 mm 3.5 μm Each
13125229 100 mm 4.6 mm 3.5 μm Each
13155229 50 mm 1 mm 5 μm Each
13165229 100 mm 1 mm 5 μm Each
13175229 150 mm 1 mm 5 μm Each
13185229 50 mm 2.1 mm 5 μm Each
13195229 100 mm 2.1 mm 5 μm Each
13105239 150 mm 2.1 mm 5 μm Each
13115239 250 mm 2.1 mm 5 μm Each
13125239 50 mm 4.6 mm 5 μm Each
13135239 100 mm 4.6 mm 5 μm Each
13145239 150 mm 4.6 mm 5 μm Each
13155239 250 mm 4.6 mm 5 μm Each
13293090 100 mm 4.6 mm 10 μm Each
13203100 150 mm 4.6 mm 10 μm Each
13213100 250 mm 4.6 mm 10 μm Each
13283090 50 mm 4.6 mm 10 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
This item is not returnable. View return policy
Product Code. 13115229 Supplier Waters Corporation Supplier No. 186003611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The XBridge BEH300 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations.

  • The wide-pore (300 Å) trifunctionally bonded BEH particle offers you the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 50 mm
Max. Pressure 6000 psi (415 Bar)
Particle Shape Spherical
Particle Size 3.5 μm
Pore Size 300 Å
Quantity 1/Ea.
USP Type L1
Endcapped Yes
Type Column
For Use With (Application) Peptides
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.