Learn More
Waters XBridge Peptide BEH C18 OBD Prep Column, 300A, 10um, 10 mm X 50 mm
Brand:  Waters 186008198
Encompass
XBridge Peptide BEH C18 OBD Prep Column, 300A, 10um, 10 mm X 50 mm
Product Code. 17134482
  • € 2145.00 / Each