missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XCL2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06178MFV500
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
XCL2 Polyclonal antibody specifically detects XCL2 in Human samples. It is validated for ELISA
Tekniske data
| XCL2 | |
| Polyclonal | |
| ELISA | |
| C motif chemokine 2, chemokine (C motif) ligand 2, cytokine SCM-1 beta, SCM-1b, SCM1B, SCYC2XC chemokine ligand 2, small inducible cytokine subfamily C, member 2 | |
| Produced in rabbits immunized with purified, recombinant Human XCL2 (Uniprot#: Q9UBD3; Met1-Gly114) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 6846 | |
| Store at 4C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion