missing translation for 'onlineSavingsMsg'
Learn More
Learn More
xCT Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-318DL594
This item is not returnable.
View return policy
Description
xCT Polyclonal antibody specifically detects xCT in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation (Negative), Dual RNAscope ISH-IHC
Specifications
| xCT | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DyLight 594 | |
| Amino acid transport system xc-, Calcium channel blocker resistance protein CCBR1, CCBR1, cystine/glutamate transporter, SLC7A11, solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11, Solute carrier family 7 member 11, solute carrier family 7, (cationic amino acid transporter, y+ system) member 11, xCT | |
| This xCT Antibody was prepared from a synthetic peptide made to an N-terminal region of the human xCT protein (between residues 1-50) [UniProt Q9UPY5]. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Cell Biology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
| 23657 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction