missing translation for 'onlineSavingsMsg'
Learn More
Learn More
xCT Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-318C
This item is not returnable.
View return policy
Description
xCT Polyclonal specifically detects xCT in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| xCT | |
| Polyclonal | |
| Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 5 μg/mL | |
| Q9UPY5 | |
| SLC7A11 | |
| This xCT Antibody was prepared from a synthetic peptide made to an N-terminal region of the human xCT protein (between residues 1-50) [UniProt Q9UPY5]. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse and rat (PMID 21540084). | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Amino acid transport system xc-, Calcium channel blocker resistance protein CCBR1, cystine/glutamate transporter, Member 11 (Slc7a11), Solute carrier family 7 member 11, solute carrier family 7, (cationic amino acid transporter, y+ system) member 11, xCTCCBR1 | |
| Rabbit | |
| 55 kDa | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Cell Biology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
| 23657 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction