missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ xCT Antibody (SLC7A11/9136R) [DyLight 755], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27052IR
This item is not returnable.
View return policy
Description
xCT Monoclonal antibody specifically detects xCT in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| xCT | |
| Monoclonal | |
| DyLight 755 | |
| Amino acid transport system xc-, Calcium channel blocker resistance protein CCBR1, CCBR1, cystine/glutamate transporter, SLC7A11, solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11, Solute carrier family 7 member 11, solute carrier family 7, (cationic amino acid transporter, y+ system) member 11, xCT | |
| Recombinant fragments of human xCT protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Cell Biology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
| 23657 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| SLC7A11/9136R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction