missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XDH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580235
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat liver tissue, mouse liver tissue. IHC: mouse intestine tissue, rat intestine tissue. Flow: A431 cell.
XDH belongs to the group of molybdenum-containing hydroxylases involved in the oxidative metabolism of purines. The enzyme is a homodimer. This protein can be converted to xanthine oxidase by reversible sulfhydryl oxidation or by irreversible proteolytic modification. Defects in xanthine dehydrogenase cause xanthinuria, may contribute to adult respiratory stress syndrome, and may potentiate influenza infection through an oxygen metabolite-dependent mechanism.
Specifications
| XDH | |
| Polyclonal | |
| Unconjugated | |
| XDH | |
| Unknown (protein for MGC:127098); xanthine dehydrogenase; Xanthine dehydrogenase/oxidase; xanthine oxidase; Xanthine oxidoreductase; XD; Xdh; xdh antibody; XDHA; xdha antibody; XO; XOR; xor antibody; Xox1; Xox-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22436, 497811, 7498 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P22985, P47989, Q00519 | |
| XDH | |
| E. coli-derived human Xanthine Oxidase recombinant protein (Position: T2-K343). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction