missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XG Polyclonal Antibody

Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | XG |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | XG |
| Gene Accession No. | P55808 |
| Gene Alias | Glycoprotein Xg; MGC118758; MGC118759; MGC118760; MGC118761; PBDX; Protein PBDX; XG; Xg blood group; Xg blood group (pseudoautosomal boundary-divided on the X chromosome); XG glycoprotein; XGPY |
| Gene Symbols | XG |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?