Learn More
Ximelagatran Nitrile, TRC
Specifications
Specifications
| Chemical Name or Material | Ximelagatran Nitrile |
| CAS | 260790-61-2 |
| Molecular Formula | C24H32N4O4 |
| Quantity | 1 mg |
| Synonym | N-[(1R)-2-[(2S)-2-[[[(4-Cyanophenyl)methyl]amino]carbonyl]-1-azetidinyl]-1-cyclohexyl-2-oxoethyl]glycine Ethyl Ester |
| InChI Formula | InChI=1S/C24H32N4O4/c1-2-32-21(29)16-26-22(19-6-4-3-5-7-19)24(31)28-13-12-20(28)23(30)27-15-18-10-8-17(14-25)9-11-18/h8-11,19-20,22,26H,2-7,12-13,15-16H2,1H3,(H,27,30)/t20-,22+/m0/s1 |
| SMILES | CCOC(=O)CN[C@H](C1CCCCC1)C(=O)N2CC[C@H]2C(=O)NCc3ccc(cc3)C#N |
| IUPAC Name | ethyl 2-[[(1R)-2-[(2S)-2-[(4-cyanophenyl)methylcarbamoyl]azetidin-1-yl]-1-cyclohexyl-2-oxoethyl]amino]acetate |
| Molecular Weight (g/mol) | 440.54 |
| Formula Weight | 440.2424 |