Learn More
Ximelagatran, TRC
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Specifications
Specifications
| Chemical Name or Material | Ximelagatran |
| CAS | 192939-46-1 |
| Molecular Formula | C24 H35 N5 O5 |
| Quantity | 0.5 mg |
| Synonym | Glycine','N-[(1R)-1-cyclohexyl-2-[(2S)-2-[[[[4-[(hydroxyamino)iminomethyl]phenyl]methyl]amino]carbonyl]-1-azetidinyl]-2-oxoethyl]-','ethyl ester, Exanta, H 376/95, Ximelagatran |
| InChI Formula | InChI=1S/C24H35N5O5/c1-2-34-20(30)15-26-21(17-6-4-3-5-7-17)24(32)29-13-12-19(29)23(31)27-14-16-8-10-18(11-9-16)22(25)28-33/h8-11,17,19,21,26,33H,2-7,12-15H2,1H3,(H2,25,28)(H,27,31)/t19-,21+/m0/s1 |
| SMILES | CCOC(=O)CN[C@H](C1CCCCC1)C(=O)N2CC[C@H]2C(=O)NCc3ccc(cc3)C(=N)NO |
| IUPAC Name | ethyl 2-[[(1R)-1-cyclohexyl-2-[(2S)-2-[[4-(N-hydroxycarbamimidoyl)phenyl]methylcarbamoyl]azetidin-1-yl]-2-oxoethyl]amino]acetate |
| Molecular Weight (g/mol) | 473.57 |
| Formula Weight | 473.2638 |