missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XIRP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548605
This item is not returnable.
View return policy
Description
XIRP1 Polyclonal Antibody for Western Blot, IHC (P)
Protects actin filaments from depolymerization.
Specifications
| XIRP1 | |
| Polyclonal | |
| Unconjugated | |
| XIRP1 | |
| AI415219; cardiomyopathy associated 1; cardiomyopathy-associated protein 1; CMYA1; LOC100910104; mXin alpha; Xin; xin actin binding repeat containing 1; xin actin-binding repeat containing 1; xin actin-binding repeat-containing protein 1; xin actin-binding repeat-containing protein 1-like; Xin alpha; XIRP1 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 165904 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q702N8 | |
| XIRP1 | |
| KLH conjugated synthetic peptide between 1338-1367 amino acids from the C-terminal region of human XIRP1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction