missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ XK Polyclonal Antibody
GREENER_CHOICE

Product Code. 16547731
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16547731 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16547731 Supplier Invitrogen™ Supplier No. PA590227

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: mouse pancreas, rat pancreas; Cellular Location: Membrane, Multi-pass membrane protein.

The XK protein is a membrane-bound protein associated with the endoplasmic reticulum (ER) and forms part of the ER membrane complex (EMC), which is critical for various cellular processes. This protein is involved in the insertion, folding, and assembly of membrane proteins in eukaryotic cells. The XK protein's role in the ER highlights its importance in maintaining proper cell function, as the ER is integral in protein synthesis, lipid metabolism, and calcium storage. Furthermore, the XK protein's involvement in these processes underscores its significance in cellular homeostasis and its potential contribution to pathologies when mutated or dysfunctional.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XK
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.00 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene XK
Gene Accession No. P51811, Q5GH61, Q9QXY7
Gene Alias 1810038K19Rik; Kell blood group precursor (McLeod phenotype); Kell blood group precursor (McLeod phenotype) homolog; kell complex 37 kDa component; KX; Kx antigen; MCLDS; McLeod syndrome gene homolog; membrane transport protein XK; NA; NAC; neuroacanthocytosis; neurocanthocytosis; X1k; Xk; XK homolog; XK, Kell blood group complex subunit (McLeod syndrome); Xkh; XKR1; XK-related protein 1; X-linked Kx blood group; X-linked Kx blood group (McLeod syndrome); X-linked Kx blood group (McLeod syndrome) homolog; Xrg1
Gene Symbols XK
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22439, 497078, 7504
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.