missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XKR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557147
This item is not returnable.
View return policy
Description
Immunogen sequence: METVFEEMDE ESTGGVSSSK EEIVLGQRLH Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 38%.
XKRX (MIM 300684) and XKR3 are homologs of the Kell blood group precursor XK (MIM 314850), which is a putative membrane transporter and a component of the XK/Kell complex of the Kell blood group system (Calenda et al., 2006 [PubMed 16431037]).[supplied by OMIM, Mar 2008].
Specifications
| XKR3 | |
| Polyclonal | |
| Unconjugated | |
| XKR3 | |
| expressed in testis; X Kell blood group precursor-related family, member 3; x Kell blood group-related 3; XK related 3; XK, Kell blood group complex subunit-related family, member 3; XKR3; XK-related protein 3; X-linked Kx blood group related 3; XRG3; XTES | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 150165 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5GH77 | |
| XKR3 | |
| Recombinant protein corresponding to Human XKR3. Recombinant protein control fragment (Product #RP-95371). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction