missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XLF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2258
This item is not returnable.
View return policy
Description
XLF Polyclonal antibody specifically detects XLF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| XLF | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA | |
| Cernunnos, non-homologous end-joining factor 1, nonhomologous end-joining factor 1, Protein cernunnos, XLFFLJ12610, XRCC4-like factor | |
| The immunogen recognized by this antibody maps to a region between residue 250 and the C-terminus (residue 299) of human XRCC4-Like Factor using the numbering given in entry NP_079058.1 (GeneID 79840). | |
| 0.1 mL | |
| Cancer, Chromatin Research, DNA Repair | |
| 79840 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:2000 -1:10000, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 2 -5 μg/mg lysate, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| NP_079058.1 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction