missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ XMU MP 1
GREENER_CHOICE

Product Code. 18787108
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18787108 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18787108 Supplier Tocris Bioscience™ Supplier No. 6482/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective MST1/2 inhibitor

XMU MP 1 is a potent and selective ATP-competitive mammalian sterile 20-like kinase (MST) 1/2 inhibitor (IC50 values are 38.1 and 71.1 nM for MST2 and MST1, respectively). Enhances activity of downstream YAP in vitro. Augments mouse intestinal repair and promotes liver repair and regeneration in a mouse model of acute and chronic liver injury. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[(6,10-Dihydro-5,10-dimethyl-6-oxo-5H-pyrimido[5,4-b]thieno[3,2-e][1,4]diazepin-2-yl)amino]benzenesulfonamide
CAS 2061980-01-4
Quantity 10 mg
Molecular Formula C17H16N6O3S2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.