missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XP32 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06224PE
This item is not returnable.
View return policy
Description
XP32 Polyclonal antibody specifically detects XP32 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| XP32 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| C1orf68, chromosome 1 open reading frame 68, LEP7, skin-specific protein (xp32), skin-specific protein 32, XP32 | |
| Produced in rabbits immunized with E. coli-derived Human XP32 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 100129271 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction