missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPA Antibody (2A4), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50600R
This item is not returnable.
View return policy
Description
XPA Monoclonal antibody specifically detects XPA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| XPA | |
| Monoclonal | |
| DyLight 550 | |
| DNA repair protein complementing XP-A cells, excision repair-controlling, Xeroderma pigmentosum group A-complementing protein, xeroderma pigmentosum, complementation group A, XPACXP1 | |
| His-tagged recombinant human XPA protein. | |
| 0.1 mL | |
| DNA Repair, Nucleotide Excision Repair | |
| 7507 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 2A4 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction