missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPB Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-61060AF405
This item is not returnable.
View return policy
Description
XPB Polyclonal antibody specifically detects XPB in Human, Mouse, Virus samples. It is validated for Western Blot
Specifications
| XPB | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Virus | |
| Purified |
| Western Blot | |
| Alexa Fluor 405 | |
| Basic transcription factor 2 89 kDa subunit, BTF2, BTF2 p89, DNA excision repair protein ERCC-3, DNA repair protein complementing XP-B cells, EC 3.6.1, EC 3.6.4.12, excision repair cross-complementing rodent repair deficiency, complementationgroup 3 (xeroderma pigmentosum group B complementing), GTF2H, RAD25, TFIIH, TFIIH basal transcription factor complex 89 kDa subunit, TFIIH basal transcription factor complex helicase XPB subunit, TFIIH p89, Xeroderma pigmentosum group B-complementing protein, xeroderma pigmentosum, complementation group B, XPBC, XPBTFIIH 89 kDa subunit | |
| The immunogen recognized by this antibody maps to a region between residue 732 and 782 of human excision repair cross-complementing rodent repair deficiency, complementation group 3 using the numbering given in entry NP_000113.1 (GeneID 2071). | |
| 0.1 mL | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2071 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction