missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XPD Recombinant Rabbit Monoclonal Antibody (23GB1675), Invitrogen™

Description
XPD is involved with the nucleotide excision repair pathway that is a mechanism to repair damage to DNA. The protein encoded by this gene is involved in transcription-coupled nucleotide excision repair and is an integral member of the basal transcription factor BTF2/TFIIH complex. The gene product has ATP-dependent DNA helicase activity and belongs to the RAD3/XPD subfamily of helicases. Defects in this gene can result in three different disorders, the cancer-prone syndrome xeroderma pigmentosum complementation group D, trichothiodystrophy, and Cockayne syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Antigen | XPD |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB1675 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ERCC2 |
| Gene Accession No. | P18074 |
| Gene Alias | AA407812; AU020867; AW240756; basic transcription factor 2 80 kDa subunit; BTF2 p80; COFS2; CXPD; DNA excision repair protein ERCC-2; DNA repair protein complementing XP-D cells; EM9; ERCC excision repair 2, TFIIH core complex helicase subunit; Ercc2; Ercc-2; excision repair cross-complementation group 2; excision repair cross-complementing rodent repair deficiency, complementation group 2; General transcription and DNA repair factor IIH helicase subunit XPD; MGC102762; MGC126218; MGC126219; TFIIH; TFIIH 80 kDa subunit; TFIIH basal transcription factor complex 80 kDa subunit; TFIIH basal transcription factor complex helicase subunit; TFIIH basal transcription factor complex helicase XPB subunit; TFIIH basal transcription factor complex helicase XPD subunit; TFIIH p80; TFIIH subunit XPD; TTD; TTD1; xeroderma pigmentosum complementary group D; xeroderma pigmentosum group D-complementing protein; XPD; XPDC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?