missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPF Antibody (OTI3H7) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74882
This item is not returnable.
View return policy
Description
XPF Monoclonal specifically detects XPF in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| XPF | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:1000, Flow Cytometry 1:100, CyTOF-ready | |
| DNA excision repair protein ERCC-4, DNA repair protein complementing XP-F cells, EC 3.1, ERCC11, excision repair cross-complementing rodent repair deficiency, complementationgroup 4, xeroderma pigmentosum, complementation group F, XPFcomplementing defective, in Chinese hamster | |
| Mouse | |
| 104.3 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, CyTOF | |
| OTI3H7 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| ERCC4 | |
| Full length human recombinant protein of human ERCC4(NP_005227) produced in HEK293T cell. | |
| Affinity Purified | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2072 | |
| Human, Mouse | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction