missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XPNPEP1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547427
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) XPNPEP2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes the cytosolic form of a metalloaminopeptidase that catalyzes the cleavage of the N-terminal amino acid adjacent to a proline residue. The gene product may play a role in degradation and maturation of tachykinins, neuropeptides, and peptide hormones. Alternative splicing results in multiple transcript variants.
Specifications
| XPNPEP1 | |
| Polyclonal | |
| Unconjugated | |
| Xpnpep1 | |
| Aminoacylproline aminopeptidase; aminopeptidase P, cytosolic; App; APP1; cytoplasmic aminopeptidase P; cytosolic aminopeptidase P; D230045I08Rik; OTTHUMP00000020457; OTTHUMP00000058856; RGD:621274}; sAmp; soluble aminopeptidase P; Xaa-Pro aminopeptidase 1; XPNPEP; XPNPEP1; xpnpep1 {ECO:0000312; XPNPEPL; XPNPEPL1; X-Pro aminopeptidase 1; X-Pro aminopeptidase 1, soluble; X-prolyl aminopeptidase (aminopeptidase P) 1, soluble; X-prolyl aminopeptidase 1; X-prolyl aminopeptidase 1, soluble | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 7511 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9NQW7 | |
| Xpnpep1 | |
| E. coli-derived recombinant human Aminopeptidase P1/XPNPEP1 Met1-His623. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction