missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XPO5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556293
This item is not returnable.
View return policy
Description
Immunogen sequence: MSFCVYSILG VVKRTCWPTD LEEAKAGGFV VGYTSSGNPI FRNPCTEQIL KLLDNLLALI RTHNTLYAPE MLAKMAEPFT KALDMLDAEK SAILGLPQPL LELNDSPVFK TVLERMQRFF STLYENC Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments. Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments.
Specifications
| XPO5 | |
| Polyclonal | |
| Unconjugated | |
| XPO5 | |
| 2410004H11Rik; 2700038C24Rik; AI648907; AW549301; exp5; exportin 5; exportin-5; FLJ14239; FLJ32057; FLJ45606; KIAA1291; mKIAA1291; Ran-binding protein 21; RanBp21; Xpo5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57510, 72322 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q924C1, Q9HAV4 | |
| XPO5 | |
| Recombinant protein corresponding to Human XPO5. Recombinant protein control fragment (Product #RP-91842). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction