missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ XPO5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15732871 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15732871 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15732871 Supplier Invitrogen™ Supplier No. PA554941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ASLVHLAFQI YEALRPRYLE IRAVMEQIPE IQKDSLDQFD CKLLNPSLQK VADKRRKDQF KRLIAGCIGK PLGEQFRKEV H Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments. Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPO5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene XPO5
Gene Accession No. Q924C1, Q9HAV4
Gene Alias 2410004H11Rik; 2700038C24Rik; AI648907; AW549301; exp5; exportin 5; exportin-5; FLJ14239; FLJ32057; FLJ45606; KIAA1291; mKIAA1291; Ran-binding protein 21; RanBp21; Xpo5
Gene Symbols XPO5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human XPO5. Recombinant protein control fragment (Product #RP-91844).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363194, 57510, 72322
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.