missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XPO5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578545
This item is not returnable.
View return policy
Description
Positive Control: 293T, A431, HeLa, HepG2, K562 Predicted Reactivity: Mouse (86%), Rat (86%), Chicken (80%), Rhesus Monkey (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments. Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments.
Specifications
| XPO5 | |
| Polyclonal | |
| Unconjugated | |
| XPO5 | |
| 2410004H11Rik; 2700038C24Rik; AI648907; AW549301; exp5; exportin 5; exportin-5; FLJ14239; FLJ32057; FLJ45606; KIAA1291; mKIAA1291; Ran-binding protein 21; RanBp21; Xpo5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57510 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1.59 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9HAV4 | |
| XPO5 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human Exportin 5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Amphibian | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction