missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ XPO5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16588431
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16588431 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16588431 Supplier Invitrogen™ Supplier No. PA593224

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MRLSQKWQVI NQRSLLCGED EAADENPESQ EMLEEQLVRM LTREVMDLIT VCCVSKKGAD HSSAPPADGD DEEMMATEVT PSAMAELTDL GKCLMKHEDV CTALLITAFN SLAWKDTLSC QRTTSQLCWP LLKQVLSGTL LADAVTWLFT SVLKGLQMHG QHDGCMASLV HLAFQIYEAL RPRYLEIRAV MEQIPEIQKD SLDQFDCKLL NPSLQKVADK RRKDQFKRLI AGCIGKPLGE QFRKEVHIKN LPSLFKKTKP MLETEVLDND GGGLATIFEP; Positive Samples: MCF7, U-251MG, NIH/3T3, HeLa, PC-12, Mouse brain, Rat brain; Cellular Location: Cytoplasm, Nucleus.

Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments. Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPO5
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 10.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene XPO5
Gene Accession No. Q924C1, Q9HAV4
Gene Alias 2410004H11Rik; 2700038C24Rik; AI648907; AW549301; exp5; exportin 5; exportin-5; FLJ14239; FLJ32057; FLJ45606; KIAA1291; mKIAA1291; Ran-binding protein 21; RanBp21; Xpo5
Gene Symbols XPO5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 925-1204 of human Exportin 5 (Exportin 5 (XPO5)) (NP_0658011).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363194, 57510, 72322
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.