missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPO6 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-59853AF350
This item is not returnable.
View return policy
Description
XPO6 Polyclonal antibody specifically detects XPO6 in Human samples. It is validated for Western Blot
Specifications
| XPO6 | |
| Polyclonal | |
| Western Blot | |
| Exp6, exportin 6, exportin-6, FLJ22519, KIAA0370EXP6, RAN binding protein 20, Ran-binding protein 20, RANBP20 | |
| The immunogen recognized by this antibody maps to a region between residue 1075 and 1125 of human exportin 6 using the numbering given in entry NP_055986.1 (GeneID 23214). | |
| 0.1 mL | |
| Signal Transduction | |
| 23214 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction