missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPO6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-59853
This item is not returnable.
View return policy
Description
XPO6 Polyclonal specifically detects XPO6 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications
| XPO6 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| Exp6, exportin 6, exportin-6, FLJ22519, KIAA0370EXP6, RAN binding protein 20, Ran-binding protein 20, RANBP20 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 23214 | |
| Store at 4C. Do not freeze. |
| Immunoprecipitation | |
| 1.0 mg/mL | |
| Immunoprecipitation 2-5 ug/mg lysate | |
| Q96QU8 | |
| XPO6 | |
| The immunogen recognized by this antibody maps to a region between residue 1075 and 1125 of human exportin 6 using the numbering given in entry NP_055986.1 (GeneID 23214). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction