missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XRCC1 Antibody (OTI2F8), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74891F
This item is not returnable.
View return policy
Description
XRCC1 Monoclonal specifically detects XRCC1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| XRCC1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| XRCC1 | |
| Full length recombinant protein of human XRCC1 (NP_006288) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2F8 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DNA repair protein XRCC1, RCC, X-ray repair complementing defective repair in Chinese hamster cells 1, X-ray repair cross-complementing protein 1, X-ray-repair, complementing defective, repair in Chinese hamster | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Base Excision Repair, Cancer, DNA Repair, Homologous Recombination, Phospho Specific, Tumor Suppressors | |
| 7515.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction