missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XRCC1 Monoclonal Antibody (7B4C12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548629
This item is not returnable.
View return policy
Description
XRCC1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, ELISA
XRCC1 (X-ray cross complementing factor-1) is involved in DNA base excision repair. XRCC1 is a 70 kDa protein that has been shown to be physically associated with other DNA repair enzymes, including poly (ADP-ribose) polymerase (PARP), DNA Ligase III and DNA polymerase beta. XRCC1 contains a BRCT domain (for BRCA1 carboxyl terminus) which was originally found in the tumor suppressor protein BRCA1. Recently, the BRCT domain has been shown to mediate the protein-protein interaction of XRCC1 and DNA ligase III-alpha.
Specifications
| XRCC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P18887 | |
| XRCC1 | |
| Purified recombinant fragment of human XRCC1 (AA: 1-150) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 7B4C12 | |
| Unconjugated | |
| XRCC1 | |
| DNA repair protein XRCC1; RCC; X-ray repair complementing defective repair in Chinese hamster cells 1; X-ray repair cross complementing 1; x-ray repair cross-complementing group 1 protein; X-ray repair cross-complementing protein 1; XRCC1; Xrcc-1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 7515 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction