missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ XRCC1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15905805
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15905805 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15905805 Supplier Invitrogen™ Supplier No. PA580237

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human 293T whole cell, human K562 whole cell, human Jurkat whole cell, rat brain tissue, rat liver tissue, mouse brain tissue, mouse liver tissue. IHC: rat kidney tissue, human placenta tissue, human colon cancer tissue, human mammary cancer tissue IHC-F: rat intestine tissue, human placenta tissue, mouse intestine tissue. ICC/IF: NIH3T3 cell. Flow: THP-1 cell.

XRCC1 (X-ray cross complementing factor-1) is involved in DNA base excision repair. XRCC1 is a 70 kDa protein that has been shown to be physically associated with other DNA repair enzymes, including poly (ADP-ribose) polymerase (PARP), DNA Ligase III and DNA polymerase beta. XRCC1 contains a BRCT domain (for BRCA1 carboxyl terminus) which was originally found in the tumor suppressor protein BRCA1. Recently, the BRCT domain has been shown to mediate the protein-protein interaction of XRCC1 and DNA ligase III-alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XRCC1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene XRCC1
Gene Accession No. P18887, Q60596, Q9ESZ0
Gene Alias DNA repair protein XRCC1; RCC; X-ray repair complementing defective repair in Chinese hamster cells 1; X-ray repair cross complementing 1; x-ray repair cross-complementing group 1 protein; X-ray repair cross-complementing protein 1; XRCC1; Xrcc-1
Gene Symbols XRCC1
Host Species Rabbit
Immunogen E. coli-derived human XRCC1 recombinant protein (Position: E538-A633).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22594, 7515, 84495
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.