missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XRCC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595935
This item is not returnable.
View return policy
Description
Immunogen sequence: MCSAFHRAES GTELLARLEG RSSLKEIEPN LFADEDSPVH GDILEFHGPE GTGKTEMLYH LTARCILPKS EGGLEVEVLF IDT Positive Samples: Mouse testis; Cellular Location: Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center.
The x-ray repair cross-complementing (XRCC) proteins are responsible for efficiently repairing and maintaining genetic stability following DNA base damage. These genes share sequence similarity with the yeast DNA repair protein Rad5. XRCC2 and XRCC3 are both involved in maintaining chromosome stability during cell division. XRCC2 is required for efficient repair of DNA double-strand breaks by homologous recombination between sister chromatids, and XRCC3 interacts directly with Rad51 to cooperate with Rad51 during recombinational repair.
Specifications
| XRCC2 | |
| Polyclonal | |
| Unconjugated | |
| XRCC2 | |
| 4921524O04Rik; 8030409M04Rik; DKFZp781P0919; DNA repair protein XRCC2; FANCU; RAD51; RecA; recA/RAD51 family protein; RGD1564823; X-ray repair complementing defective repair in Chinese hamster cells 2; X-ray repair cross complementing 2; X-ray repair cross complementing protein 2; X-ray repair cross-complementing protein 2; Xrcc2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 499966, 57434, 7516 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 2.03 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43543, Q9CX47 | |
| XRCC2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-83 of human XRCC2 (NP_0054221). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction