missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XRCC3 Antibody (10F1/6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08967CL1
This item is not returnable.
View return policy
Description
XRCC3 Monoclonal antibody specifically detects XRCC3 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
| XRCC3 | |
| Monoclonal | |
| CoraFluor 1 | |
| CMM6, DNA repair protein XRCC3, X-ray repair complementing defective repair in Chinese hamster cells 3, X-ray repair cross-complementing protein 3 | |
| Recombinant full-length human XRCC3 protein (Uniprot: O43542) | |
| 0.1 mL | |
| Apoptosis, Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents, Homologous Recombination, Tumor Suppressors | |
| 7517 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 10F1/6 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction