missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XRCC3 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-165B
This item is not returnable.
View return policy
Description
XRCC3 Polyclonal specifically detects XRCC3 in Human, Drosophila, Hamster samples. It is validated for Western Blot.
Specifications
| XRCC3 | |
| Polyclonal | |
| Western Blot 1:5000 | |
| O43542 | |
| XRCC3 | |
| A synthetic partial peptide of human XRCC3. [UniProt# O43542] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Drosophila, Hamster | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CMM6, DNA repair protein XRCC3, X-ray repair complementing defective repair in Chinese hamster cells 3, X-ray repair cross-complementing protein 3 | |
| Rabbit | |
| 38 kDa | |
| 0.1 mL | |
| Apoptosis, Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents, Homologous Recombination, Tumor Suppressors | |
| 7517 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction