missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XRCC3 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98844R
This item is not returnable.
View return policy
Description
XRCC3 Polyclonal antibody specifically detects XRCC3 in Human samples. It is validated for Western Blot
Specifications
| XRCC3 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| CMM6, DNA repair protein XRCC3, X-ray repair complementing defective repair in Chinese hamster cells 3, X-ray repair cross-complementing protein 3 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human XRCC3. | |
| 0.1 mL | |
| Apoptosis, Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents, Homologous Recombination, Tumor Suppressors | |
| 7517 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction