Learn More
Invitrogen™ XRCC6BP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588836
Description
Immunogen sequence: MAGAPDERRR GPAAGEQLQQ QHVSCQVFPE RLAQGNPQQG FFSSFFTSNQ KCQLRLLKTL ETNPYVKLLL DAMKHSGCAV NKDRHFSCED CNGNVSGGFD ASTSQIVLCQ NNIHNQAHMN RVVTHELIHA FDHCRAHVDW FTNIRHLACS EVRAANLSGD CSLVNEIFRL HFGLKQHHQT CVRDRATLSI LAVRNISKEV AKKAVDEVFE SCFNDHEPFG RIPHNKTYAR YAHRDFENRD RYYSNI; Positive Samples: Mouse brain, Rat brain.
The protein encoded by this gene is amplified in glioblastomas and interacts with the DNA binding subunit of DNA-dependent protein kinase. This kinase is involved in double-strand break repair (DSB), and higher expression of the encoded protein increases the efficiency of DSB. In addition, comparison to orthologous proteins strongly suggests that this protein is a metalloprotease important in the biosynthesis of mitochondrial ATPase. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| XRCC6BP1 | |
| Polyclonal | |
| Unconjugated | |
| ATP23 | |
| 1110068E08Rik; 2410012H02Rik; atp23; ATP23 metallopeptidase and ATP synthase assembly factor homolog; Ku70 binding protein 3; Ku70-binding protein 3; KUB3; LOW QUALITY PROTEIN: mitochondrial inner membrane protease ATP23 homolog; mitochondrial inner membrane protease ATP23 homolog; XRCC6 binding protein 1; XRCC6-binding protein 1; Xrcc6bp1; zgc:162885 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 299828, 68876 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9CWQ3 | |
| ATP23 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-246 of human ATP23 (NP_1505921). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.