missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XTP3TPA Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18474AF488
This item is not returnable.
View return policy
Description
XTP3TPA Polyclonal antibody specifically detects XTP3TPA in Human, Mouse samples. It is validated for Western Blot
Specifications
| XTP3TPA | |
| Polyclonal | |
| Western Blot | |
| dCTP pyrophosphatase 1, dCTPase 1, Deoxycytidine-triphosphatase 1, EC 3.6.1.12, MGC5627, RS21C6XTP3-transactivated gene A protein, XTP3TPACDA03, XTP3-transactivated protein A | |
| Recognizes region between residues Between between 120 and 170.. | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 79077 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction