missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XTP3TPA Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18473AF594
This item is not returnable.
View return policy
Description
XTP3TPA Polyclonal antibody specifically detects XTP3TPA in Human samples. It is validated for Western Blot
Specifications
| XTP3TPA | |
| Polyclonal | |
| Western Blot | |
| dCTP pyrophosphatase 1, dCTPase 1, Deoxycytidine-triphosphatase 1, EC 3.6.1.12, MGC5627, RS21C6XTP3-transactivated gene A protein, XTP3TPACDA03, XTP3-transactivated protein A | |
| Recognizes region between residues Between between 120 and 170.. | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 79077 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido