missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XTP4 Antibody (OTI2D5), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74904G
This item is not returnable.
View return policy
Description
XTP4 Monoclonal specifically detects XTP4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| XTP4 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MIEN1 | |
| Full length recombinant protein of human XTP4 (NP_115715) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2D5 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| C35, chromosome 17 open reading frame 37, HBV XAg-transactivated protein 4, HBV X-transactivated gene 4 protein, MGC14832, ORB3, protein C17orf37, Protein C35, XTP4RDX12 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 84299.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction