Learn More
XY1, MedChemExpress
Product Code. 16477858
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477858 10 mg 10mg
16453807 50 mg 50mg
16410328 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16477858 Supplier Medchem Express Supplier No. HY19714
MedChemExpress XY1 is a very close analogue of SGC707 (a potent, selective, and non-competitive inhibitor of PRMT3 with IC50 of 31 nM), but XY1 is completely inactive.
XY1 is a very close analogue of SGC707 (a potent, selective, and non-competitive inhibitor of PRMT3 with IC50 of 31 nM), but XY1 is completely inactive.
Specifications
Chemical Name or Material XY1
Physical Form Solid
CAS 1624117-53-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C17H19N3O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 67.5 mg/mL (227.01 mM; Need ultrasonic)
SMILES O=C(NCC(N1CCCC1)=O)NC2=CC=C3C=CC=CC3=C2
Molecular Weight (g/mol) 297.35
Formula Weight 297.35
Percent Purity 99.03%
Grade Research
Show More Show Less
Research purposes only