Learn More
STEMCELL Technologies Y-27632 (Dihydrochloride)
Product Code. p-7217391
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
50 mg
5 x 10 mg
Unit Size:
1mg
50mg
5mg
Product Code. 15537014
Brand: STEMCELL Technologies 72308
RHO/ROCK pathway inhibitor; Inhibits ROCK1 and ROCK2
  • Y-27632 is a cell-permeable, highly potent and selective inhibitor of Rho-associated, coiled-coil containing protein kinase (ROCK)
  • Y-27632 inhibits both ROCK1 (Ki = 220 nM) and ROCK2 (Ki = 300 nM) by competing with ATP for binding to the catalytic site
Specifications
CAS 129830-38-2
Quantity 50 mg
Molecular Formula C14H21N3O · 2HCl